Anti-DAPP1 Mouse Monoclonal Antibody [clone: 716804]

Supplier: R & D Systems
MAB7024
102886-882EA 561.11 USD
102886-882
Anti-DAPP1 Mouse Monoclonal Antibody [clone: 716804]
Antibodies
Type: Primary
Antigen: DAPP1
Clonality: Monoclonal
Clone: 716804
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR