Anti-DEK Mouse Monoclonal Antibody [clone: 715511]

Supplier: R & D Systems
MAB70201
102886-880EA 561.11 USD
102886-880
Anti-DEK Mouse Monoclonal Antibody [clone: 715511]
Antibodies
Type: Primary
Antigen: DEK
Clonality: Monoclonal
Clone: 715511
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR