Anti-DEK Mouse Monoclonal Antibody [clone: 715524]

Supplier: R & D Systems
MAB7020
102886-878EA 594.37 USD
102886-878
Anti-DEK Mouse Monoclonal Antibody [clone: 715524]
Antibodies
Type: Primary
Antigen: DEK
Clonality: Monoclonal
Clone: 715524
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR