Anti-PDCD4 Mouse Monoclonal Antibody [clone: 808508]

Supplier: R & D Systems
MAB7019
102886-876EA 577.02 USD
102886-876
Anti-PDCD4 Mouse Monoclonal Antibody [clone: 808508]
Antibodies
Type: Primary
Antigen: PDCD4
Clonality: Monoclonal
Clone: 808508
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR