Anti-SSTR3 Mouse Monoclonal Antibody [clone: 576017]

Supplier: R & D Systems
MAB7018
102886-874EA 584.25 USD
102886-874
Anti-SSTR3 Mouse Monoclonal Antibody [clone: 576017]
Antibodies
Type: Primary
Antigen: SSTR3
Clonality: Monoclonal
Clone: 576017
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR