Anti-UBB Mouse Monoclonal Antibody [clone: 83406]

Supplier: R & D Systems
MAB701
220021-456EA 594.37 USD
220021-456
Anti-UBB Mouse Monoclonal Antibody [clone: 83406]
Antibodies
Type: Primary
Antigen: UBB
Clonality: Monoclonal
Clone: 83406
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR