Anti-NCK1 Mouse Monoclonal Antibody [clone: 714506]

Supplier: R & D Systems
MAB7008
102886-872EA 577.02 USD
102886-872
Anti-NCK1 Mouse Monoclonal Antibody [clone: 714506]
Antibodies
Type: Primary
Antigen: NCK1
Clonality: Monoclonal
Clone: 714506
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR