Anti-VSIR Rat Monoclonal Antibody [clone: 742002]

Supplier: R & D Systems
MAB70051
102886-868EA 594.37 USD
102886-868
Anti-VSIR Rat Monoclonal Antibody [clone: 742002]
Antibodies
Type: Primary
Antigen: VSIR
Clonality: Monoclonal
Clone: 742002
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR