Anti-GABBR1 Mouse Monoclonal Antibody [clone: 751216]

Supplier: R & D Systems
MAB7000
102886-866EA 584.25 USD
102886-866
Anti-GABBR1 Mouse Monoclonal Antibody [clone: 751216]
Antibodies
Type: Primary
Antigen Symbol: GABBR1
Antigen Name: gamma-aminobutyric acid (GABA) B receptor 1
Antigen Synonyms: -
Clonality: Monoclonal
Clone: 751216
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR