Anti-CXCR6 Mouse Monoclonal Antibody [clone: 56811]

Supplier: R & D Systems
MAB699-100
102886-862EA 633.42 USD
102886-862
Anti-CXCR6 Mouse Monoclonal Antibody [clone: 56811]
Antibodies
Type: Primary
Antigen: CXCR6
Clonality: Monoclonal
Clone: 56811
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR