Anti-FERMT2 Mouse Monoclonal Antibody [clone: 696906]

Supplier: R & D Systems
MAB6984
102886-860EA 561.11 USD
102886-860
Anti-FERMT2 Mouse Monoclonal Antibody [clone: 696906]
Antibodies
Type: Primary
Antigen Symbol: FERMT2
Antigen Name: fermitin family member 2
Antigen Synonyms: KIND2, MIG2, PLEKHC1, UNC112, UNC112B, mig-2
Clonality: Monoclonal
Clone: 696906
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR