Anti-COX4I2 Mouse Monoclonal Antibody [clone: 673803]

Supplier: R & D Systems
MAB6980
102886-858EA 577.02 USD
102886-858
Anti-COX4I2 Mouse Monoclonal Antibody [clone: 673803]
Antibodies
Type: Primary
Antigen: COX4I2
Clonality: Monoclonal
Clone: 673803
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR