Anti-MIP-III Mouse Monoclonal Antibody [clone: 76621]

Supplier: R & D Systems
MAB698
220021-454EA 763.57 USD
220021-454
Anti-MIP-III Mouse Monoclonal Antibody [clone: 76621]
Antibodies
Type: Primary
Antigen: MIP-III
Clonality: Monoclonal
Clone: 76621
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Viral
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR