Anti-CD1D Mouse Monoclonal Antibody [clone: 703335]

Supplier: R & D Systems
MAB6979
102886-856EA 594.37 USD
102886-856
Anti-CD1D Mouse Monoclonal Antibody [clone: 703335]
Antibodies
Type: Primary
Antigen: CD1D
Clonality: Monoclonal
Clone: 703335
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR