Anti-NUAK1 Mouse Monoclonal Antibody [clone: 715901]

Supplier: R & D Systems
MAB6978
102886-854EA 594.37 USD
102886-854
Anti-NUAK1 Mouse Monoclonal Antibody [clone: 715901]
Antibodies
Type: Primary
Antigen: NUAK1
Clonality: Monoclonal
Clone: 715901
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR