Anti-STIM1 Mouse Monoclonal Antibody [clone: 705129]

Supplier: R & D Systems
MAB69761
102886-850EA 577.02 USD
102886-850
Anti-STIM1 Mouse Monoclonal Antibody [clone: 705129]
Antibodies
Type: Primary
Antigen: STIM1
Clonality: Monoclonal
Clone: 705129
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR