Anti-STIM1 Mouse Monoclonal Antibody [clone: 705111]

Supplier: R & D Systems
MAB6976
102886-848EA 577.02 USD
102886-848
Anti-STIM1 Mouse Monoclonal Antibody [clone: 705111]
Antibodies
Type: Primary
Antigen: STIM1
Clonality: Monoclonal
Clone: 705111
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR