Anti-ICOS Mouse Monoclonal Antibody [clone: 669222]

Supplier: R & D Systems
MAB6975
102886-846EA 633.42 USD
102886-846
Anti-ICOS Mouse Monoclonal Antibody [clone: 669222]
Antibodies
Type: Primary
Antigen: ICOS
Clonality: Monoclonal
Clone: 669222
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR