Anti-CLDN19 Mouse Monoclonal Antibody [clone: 693202]

Supplier: R & D Systems
MAB6970
102886-844EA 594.37 USD
102886-844
Anti-CLDN19 Mouse Monoclonal Antibody [clone: 693202]
Antibodies
Type: Primary
Antigen: CLDN19
Clonality: Monoclonal
Clone: 693202
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR