Anti-DMPK Mouse Monoclonal Antibody [clone: 713931]

Supplier: R & D Systems
MAB6967
102886-840EA 594.37 USD
102886-840
Anti-DMPK Mouse Monoclonal Antibody [clone: 713931]
Antibodies
Type: Primary
Antigen: DMPK
Clonality: Monoclonal
Clone: 713931
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR