Anti-AGT Rat Monoclonal Antibody [clone: 754438]

Supplier: R & D Systems
MAB6966
102886-838EA 577.02 USD
102886-838
Anti-AGT Rat Monoclonal Antibody [clone: 754438]
Antibodies
Type: Primary
Antigen: AGT
Clonality: Monoclonal
Clone: 754438
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR