Anti-AXL Mouse Monoclonal Antibody [clone: 713610]

Supplier: R & D Systems
MAB6965
102886-836EA 623.29 USD
102886-836
Anti-AXL Mouse Monoclonal Antibody [clone: 713610]
Antibodies
Simple Western

Type: Primary
Antigen: AXL
Clonality: Monoclonal
Clone: 713610
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR