Anti-IL1B Mouse Monoclonal Antibody [clone: 615417]

Supplier: R & D Systems
MAB6964
102886-834EA 600.15 USD
102886-834
Anti-IL1B Mouse Monoclonal Antibody [clone: 615417]
Antibodies
Simple Western

Type: Primary
Antigen: IL1B
Clonality: Monoclonal
Clone: 615417
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR