Anti-CCI Mouse Monoclonal Antibody [clone: 112803]

Supplier: R & D Systems
MAB6961
220021-453EA 763.57 USD
220021-453
Anti-CCI Mouse Monoclonal Antibody [clone: 112803]
Antibodies
Type: Primary
Antigen: CCI
Clonality: Monoclonal
Clone: 112803
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Viral
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR