Anti-CCI Mouse Monoclonal Antibody [clone: 112811]

Supplier: R & D Systems
MAB696
220021-452EA 763.57 USD
220021-452
Anti-CCI Mouse Monoclonal Antibody [clone: 112811]
Antibodies
Type: Primary
Antigen: CCI
Clonality: Monoclonal
Clone: 112811
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Viral
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR