Anti-EGLN3 Mouse Monoclonal Antibody [clone: 700210]

Supplier: R & D Systems
MAB6954
102886-832EA 566.89 USD
102886-832
Anti-EGLN3 Mouse Monoclonal Antibody [clone: 700210]
Antibodies
Type: Primary
Antigen: EGLN3
Clonality: Monoclonal
Clone: 700210
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR