Anti-XCL1 Mouse Monoclonal Antibody [clone: 109001]

Supplier: R & D Systems
MAB6951
220021-451EA 772.25 USD
220021-451
Anti-XCL1 Mouse Monoclonal Antibody [clone: 109001]
Antibodies
Type: Primary
Antigen: XCL1
Clonality: Monoclonal
Clone: 109001
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR