Anti-ITGAX Hamster Monoclonal Antibody [clone: N418]

Supplier: R & D Systems
MAB69501
102886-830EA 594.37 USD
102886-830
Anti-ITGAX Hamster Monoclonal Antibody [clone: N418]
Antibodies
Type: Primary
Antigen: ITGAX
Clonality: Monoclonal
Clone: N418
Conjugation: Unconjugated
Epitope:
Host: Hamster
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR