Anti-ITGAX Rat Monoclonal Antibody [clone: 435421]

Supplier: R & D Systems
MAB6950
102886-828EA 594.37 USD
102886-828
Anti-ITGAX Rat Monoclonal Antibody [clone: 435421]
Antibodies
Type: Primary
Antigen: ITGAX
Clonality: Monoclonal
Clone: 435421
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR