Anti-TNFSF18 Mouse Monoclonal Antibody [clone: 109101]

Supplier: R & D Systems
MAB6941
220021-449EA 772.25 USD
220021-449
Anti-TNFSF18 Mouse Monoclonal Antibody [clone: 109101]
Antibodies
Blockade of Receptor-ligand Interaction

Type: Primary
Antigen: TNFSF18
Clonality: Monoclonal
Clone: 109101
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR