Anti-IL4 Mouse Monoclonal Antibody [clone: 855824]

Supplier: R & D Systems
MAB6939
102886-824EA 594.37 USD
102886-824
Anti-IL4 Mouse Monoclonal Antibody [clone: 855824]
Antibodies
Type: Primary
Antigen: IL4
Clonality: Monoclonal
Clone: 855824
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR