Anti-IL10 Mouse Monoclonal Antibody [clone: 262715]

Supplier: R & D Systems
MAB6932
220021-447EA 561.11 USD
220021-447
Anti-IL10 Mouse Monoclonal Antibody [clone: 262715]
Antibodies
Type: Primary
Antigen: IL10
Clonality: Monoclonal
Clone: 262715
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR