Anti-IL10 Mouse Monoclonal Antibody [clone: 148801]

Supplier: R & D Systems
MAB6931
220021-446EA 772.25 USD
220021-446
Anti-IL10 Mouse Monoclonal Antibody [clone: 148801]
Antibodies
Type: Primary
Antigen: IL10
Clonality: Monoclonal
Clone: 148801
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR