Anti-STMN2 Mouse Monoclonal Antibody [clone: 684433]

Supplier: R & D Systems
MAB6930
102886-820EA 600.15 USD
102886-820
Anti-STMN2 Mouse Monoclonal Antibody [clone: 684433]
Antibodies
Type: Primary
Antigen: STMN2
Clonality: Monoclonal
Clone: 684433
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR