Anti-IL10 Mouse Monoclonal Antibody [clone: 148806]

Supplier: R & D Systems
MAB693
220021-445EA 772.25 USD
220021-445
Anti-IL10 Mouse Monoclonal Antibody [clone: 148806]
Antibodies
Type: Primary
Antigen: IL10
Clonality: Monoclonal
Clone: 148806
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR