Anti-THEMIS Mouse Monoclonal Antibody [clone: 701010]

Supplier: R & D Systems
MAB6929
102886-818EA 561.11 USD
102886-818
Anti-THEMIS Mouse Monoclonal Antibody [clone: 701010]
Antibodies
Type: Primary
Antigen: THEMIS
Clonality: Monoclonal
Clone: 701010
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR