Anti-TCF12 Mouse Monoclonal Antibody [clone: 690129]

Supplier: R & D Systems
MAB69281
102886-816EA 561.11 USD
102886-816
Anti-TCF12 Mouse Monoclonal Antibody [clone: 690129]
Antibodies
Type: Primary
Antigen: TCF12
Clonality: Monoclonal
Clone: 690129
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR