Anti-TCF12 Mouse Monoclonal Antibody [clone: 690124]

Supplier: R & D Systems
MAB6928
102886-814EA 566.89 USD
102886-814
Anti-TCF12 Mouse Monoclonal Antibody [clone: 690124]
Antibodies
Type: Primary
Antigen: TCF12
Clonality: Monoclonal
Clone: 690124
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR