Anti-LPP Mouse Monoclonal Antibody [clone: 691121]

Supplier: R & D Systems
MAB6927
102886-812EA 561.11 USD
102886-812
Anti-LPP Mouse Monoclonal Antibody [clone: 691121]
Antibodies
Type: Primary
Antigen: LPP
Clonality: Monoclonal
Clone: 691121
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR