Anti-EPOR Mouse Monoclonal Antibody [clone: 690710]

Supplier: R & D Systems
MAB6926
102886-810EA 594.37 USD
102886-810
Anti-EPOR Mouse Monoclonal Antibody [clone: 690710]
Antibodies
Simple Western

Type: Primary
Antigen: EPOR
Clonality: Monoclonal
Clone: 690710
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR