Anti-TNS4 Mouse Monoclonal Antibody [clone: 684524]

Supplier: R & D Systems
MAB6925
102886-768EA 594.37 USD
102886-768
Anti-TNS4 Mouse Monoclonal Antibody [clone: 684524]
Antibodies
Simple Western

Type: Primary
Antigen: TNS4
Clonality: Monoclonal
Clone: 684524
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR