Anti-CD79A Mouse Monoclonal Antibody [clone: 706962]

Supplier: R & D Systems
MAB6920
102886-764EA 577.02 USD
102886-764
Anti-CD79A Mouse Monoclonal Antibody [clone: 706962]
Antibodies
Type: Primary
Antigen: CD79A
Clonality: Monoclonal
Clone: 706962
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR