Anti-TNFRSF11A Rat Monoclonal Antibody [clone: 100024]

Supplier: R & D Systems
MAB692
220021-444EA 763.57 USD
220021-444
Anti-TNFRSF11A Rat Monoclonal Antibody [clone: 100024]
Antibodies
Type: Primary
Antigen: TNFRSF11A
Clonality: Monoclonal
Clone: 100024
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR