Anti-LRRC4B Mouse Monoclonal Antibody [clone: 702008]

Supplier: R & D Systems
MAB69191
102886-762EA 594.37 USD
102886-762
Anti-LRRC4B Mouse Monoclonal Antibody [clone: 702008]
Antibodies
Type: Primary
Antigen: LRRC4B
Clonality: Monoclonal
Clone: 702008
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR