Anti-LRRC4B Mouse Monoclonal Antibody [clone: 702021]

Supplier: R & D Systems
MAB6919
102886-760EA 577.02 USD
102886-760
Anti-LRRC4B Mouse Monoclonal Antibody [clone: 702021]
Antibodies
Type: Primary
Antigen: LRRC4B
Clonality: Monoclonal
Clone: 702021
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR