Anti-PLET1 Rat Monoclonal Antibody [clone: 700738]

Supplier: R & D Systems
MAB6917
102886-756EA 577.02 USD
102886-756
Anti-PLET1 Rat Monoclonal Antibody [clone: 700738]
Antibodies
Type: Primary
Antigen: PLET1
Clonality: Monoclonal
Clone: 700738
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR