Anti-SMURF2 Mouse Monoclonal Antibody [clone: 690638]

Supplier: R & D Systems
MAB6916
102886-754EA 633.42 USD
102886-754
Anti-SMURF2 Mouse Monoclonal Antibody [clone: 690638]
Antibodies
Type: Primary
Antigen: SMURF2
Clonality: Monoclonal
Clone: 690638
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR