Anti-SH2B1 Mouse Monoclonal Antibody [clone: 700320]

Supplier: R & D Systems
MAB6915
102886-752EA 561.11 USD
102886-752
Anti-SH2B1 Mouse Monoclonal Antibody [clone: 700320]
Antibodies
Type: Primary
Antigen: SH2B1
Clonality: Monoclonal
Clone: 700320
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR