Anti-PPM1L Mouse Monoclonal Antibody [clone: 700612]

Supplier: R & D Systems
MAB6914
102886-750EA 577.02 USD
102886-750
Anti-PPM1L Mouse Monoclonal Antibody [clone: 700612]
Antibodies
Type: Primary
Antigen: PPM1L
Clonality: Monoclonal
Clone: 700612
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR