Anti-CRKL Mouse Monoclonal Antibody [clone: 668915]

Supplier: R & D Systems
MAB6910
102886-748EA 616.06 USD
102886-748
Anti-CRKL Mouse Monoclonal Antibody [clone: 668915]
Antibodies
Type: Primary
Antigen: CRKL
Clonality: Monoclonal
Clone: 668915
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR